Learn More
Mitegen ANDERSON-EVANS POLYOXOTUNG 5MG
Supplier: Mitegen MXTEW5
Anderson-Evans polyoxotungstate, 5 mg.The Anderson−Evans polyoxotungstate [TeW6O24]6− (TEW) is a universal and flexible additive for protein crystallization. With its planar structure and high negative charge, it has proven to promote crystal contacts, improve crystal quality and further provides a valuable anomalous signal for phasing due to 6 tungsten atoms[1].
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.