Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anatomical Chart Series - The Female Reproductive System Charts/Laminated Posters
SCI_ED

Shows ovary, fallopian tube, uterus and vagina.

Supplier:  Anatomical Chart Company 9781587790201

Catalog No. S96924

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Illustrates ovum, the female pelvic organs (sagittal section) and the female perineum
  • Provides views of ovary and uterus during the menstural cycle

Order Info

Size: 20W x 26 in. H.

Specifications

Specifications

Female Reproductive System Poster
Science
66.04 cm
50.8 cm
Laminated Paper
26 in.
20 in.
Views of ovary fallopian tube, uterus, vagina, ovum, the female pelvic organs (sagittal section), the female perineum, also provides views of the ovary and uterus during the menstrual cycle, 2 eyelets for easy hanging
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit