Learn More
New England Biolabs, Inc. AMV Reverse Transcriptase – 1000 units
Supplier: New England Biolabs, Inc. M0277L
Avian Myeloblastosis Virus (AMV) Reverse Transcriptase is an RNA-directed DNA polymerase. This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template.