Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Sulfate, BiotechGrade, 99%, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Ammonium Sulfate, BiotechGrade is an inorganic salt that is very soluble in water It is a widely used reagent in molecular biology and chromatography for the precipitation and fractionation of proteins, crystallization of proteins and protein-nucleic acid complexes. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications
pH | 5.0-6.0 |
CAS Max % | 100% |
Quantity | 12 kg |
Percent Purity | 99% |
Grade | Biotech |
Packaging | Poly Pail |
Residue after Ignition | 0.005% |
Appearance | White, crystallized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction