Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride, TraceSELECT™, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Quantity | 25 g |
Linear Formula | NH4Cl |
Beilstein | 4371014 |
SMILES | N.Cl |
Formula Weight | 53.49g/mol |
Percent Purity | ≥99.99% (metals basis) |
Grade | TraceSELECT™ |
Vapor Pressure | 1mmHg (160.4°C) |
Density | Vapor Density: 1.9 (vs air) |
Chemical Name or Material | Ammonium Chloride |
Safety and Handling
Hazard Statement | P305 + P351 + P338 |
Precautionary Statement | H302-H319 |
EINECSNumber | 235-186-4 |
RTECSNumber | BP4550000 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction