Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Ammonium Acetate OmniPur™ Calbiochem™,
Click to view available options
Quantity:
1 kg
5 kg
Specifications
Quantity | 5 kg |
Assay Percent Range | ≥99% |
Formula Weight | 77.1 |
Chemical Name or Material | Ammonium Acetate |
Safety and Handling
Recommended Storage | Keep container tightly closed. Keep container in a cool, well-ventilated area. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction