Learn More
Bioassay Systems EnzyChrom™ Ammonia/Ammonium Assay Kit. For quantitative determination of ammonia/ammonium concentration.
Supplier: Bioassay Systems ENH3100
For quantitative determination of ammonia/ammonium and evaluation of drug effects on their metabolism. Key Features: High sensitivity and wide linear range. Use 20 µL sample. Linear detection range 24 to 1000 µM ammonia. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of ammonia within 30 minutes. Method: OD340nm, or FL360/450nm. Samples: Serum, plasma, urine, saliva, cell culture etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 24 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.