Learn More
Hach Company Amino Acid F Reagent Solution, 100ml MDB
Supplier: Hach Company 2386432
For determination of Ultra Low Range silica by the Heteropoly Blue method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. 100 mL Marked Dropping Bottle