Learn More
United States Biological Corporation Ames Medium w/L-Glutamine (Powder), 100 L
Supplier: United States Biological Corporation A137225100L
Ames' medium was formulated to support retinal tissue in relatively short-term culture. Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central nervous system tissue in vitro.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.