Learn More
Cytiva Amersham CyDye Fluorescent Nucleotides
Ideal for multiplexing within a single sample. Cytiva Amersham CyDye Fluorescent Nucleotides are fluorescent dyes with superior characteristics for genomic and protein research. Available in multiple sizes and colors, including a multipack option for maximum versatility.
$4276.00 - $5976.00
Catalog Number | Mfr. No. | Quantity | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Description | Price | Quantity & Availability | ||||
45-200-576
|
Cytiva
PA53031 |
250 nmol | Fluorescence Color: Orange |
Each for $4,276.00
|
|
||||
45-200-578
|
Cytiva
PA55321 |
250 nmol (5 x 25 nmol Cy3-dCTP + 5 x 25 nmol Cy5-dCTP) | Fluorescence Color: Orange/Far-red |
Each for $5,976.00
|
|
||||
Description
CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. They are ideal for labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands, well suited for multiplexing within a single sample. They exhibit the following properties:- High sensitivity
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive