Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Scientific Heat Packs
SDPSCI_ED

Reusable and portable pack to produce instant heat.

Supplier:  American Scientific 322703

 View more versions of this product

Catalog No. S13164


Add to Cart

Description

Description

  • Contains super saturated solution of water and sodium acetate
  • Chain reaction created by snapping metal disc crystallize solution which gives off heat
  • Recharged back to liquid state from crystallized state by boiling in water for 10-15 minutes
  • Includes teacherapostrophes guide
  • Available in two sizes: 4 x 4 in. and 8 x 8 in.
Specifications

Specifications

Heat Pack (20.3 x 20.3cm (8 x 8 in.))
Teacher's guide
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit