Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Petzl™ America Cordex Plus Medium Weight Belay/Rappel Gloves
Click to view available options
Size:
Medium
Specifications
Certifications/Compliance | CE EN 420, CE EN 388, 4143 |
Color | Black |
Material | Leather, Neoprene |
Product Type | Extrication Gloves |
Size | Medium |
For Use With (Application) | Repelling or Belaying |
Glove Cuff Style | Hook and Loop |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction