Learn More
GFS Chemicals Inc AMCO CLEAR TURBIDITY STANDARD, 1.0 NTU | 250 ML | GFS CHEMICALS, INC. Item #: 85035 | Bulk #: 8007
Supplier: GFS Chemicals Inc 85035
AMCO CLEAR TURBIDITY STANDARD, 1.0 NTU | Amco Clear turbidity standards are traceable to NIST Primary Standards on account of their consistent particle size and photometric accuracy via UV-VIS spectroscopy. Each lot is normalized to historical lots which were formulated to dilutions of ISO17034 4000 NTU Formazin. Amco Clear turbidity standards are recognized by the US EPA as a primary stnadard for the calibration of turbitidy meters online and in the laboratory for drinking water analysis. They are safe, non-toxic, no dilutions, stable, and offer a 1 year shelf life.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.