Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plas Labs™ Amber Desiccators with Perforated Shelves
For light sensitive materials
$1451.00 - $3763.00
Catalog Number | Mfr. No. | No. of Shelves | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Shelves | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
11-389-158
|
Plas Labs™
860CGA/NS |
2 Perforated | 30.5 cm | 30.5 cm |
Each for $1,451.00
|
|
|||
11-389-161
|
Plas Labs™
861CGA/NS |
4 Perforated (2/Chamber) | 61 cm | 30.5 cm |
Each for $2,382.00
|
|
|||
11-389-164
|
Plas Labs™
862CGA/NS |
4 Perforated (2/Chamber) | 45.7 cm | 45.7 cm |
Each for $3,183.00
|
|
|||
11-389-167
|
Plas Labs™
863CGA/NS |
4 Perforated (2/Chamber) | 61 cm | 61 cm |
Each for $3,763.00
|
|
|||
Description
- Designed to protect, store, or assist in moving critical materials and prevent contamination
- Dust and moisture free enclosures useful for research specimens or sensitive electronic components
- Under positive N2 gas flow, prevent moisture from penetrating molecular structure of stored components
- Amber units are very effective for protection from harmful U.V. rays for light sensitive components
Specifications
Amber | |
Light sensitive materials |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title