Learn More
Medchemexpress LLC HY-16040 5mg Medchemexpress, AM095 (free acid) CAS:1228690-36-5 Purity:>98%
Supplier: Medchemexpress LLC HY160405MG
Medchemexpress, HY-16040 5mg AM095 (free acid) CAS:1228690-36-5 AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.