Learn More
Electron Microscopy Sciences EMS Alexander Mastoid Bone Chisel Standard, 12 mm, 6½" OAL
SDP
Supplier:  Electron Microscopy Sciences 7799002
Encompass_Preferred
Stainless steel bone chisels are used to score, cut, scrape, clean, and sculpt the bone.
Size: 12 mm
Catalog No. 50-193-1565