Learn More
United Scientific™ Aluminum Tray Dissecting
Aluminum Tray Dissecting offering an innovative way of securing the sample.
Supplier: United Scientific™ UNDISTRY04
This item is not returnable.
View return policy
Description
- Dissecting pans are made of durable aluminum with rounded corners
- The corners include holes that can be used as tie-down points to hold specimens securely during dissection
- Trays do not include wax
Specifications
Autoclavable | |
Silver Tray, Black Wax | |
Aluminum | |
14 x 10 in. |