Learn More
Perkin Elmer US LLC Aluminum Sample Pans and Covers - Qty. 100
Supplier: Perkin Elmer US LLC N5390123
The Aluminum Sample Pans and Covers are used with nonvolatile solid samples, such as polymers and pharmaceuticals. They are routinely used for applications such as polymer melting, crystallization, glass transitions of polymers, thermoplastics, and thermosets. Pans are crimped but not hermetically sealed.