Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alumina, Polishing Gamal™, Fisher Chemical
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
Specifications
Physical Form | Solid |
Quantity | 100 mL |
Color | White |
Packaging | Poly Bottle |
Boiling Point | 2980°C |
Melting Point | 2000°C |
CAS Max % | 95.44 |
Formula Weight | 101.96 |
Chemical Name or Material | Alumina, Polishing Gamal™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction