Learn More
Advalue Technologies Alumina Combustion Boat
SDP
Supplier:  Advalue Technologies AL5006
Encompass
AL-5006 Alumina Combustion Boat - Capacity 6ml - Length 75mm - Width 15mm - Height 10mm
Catalog No. NC1048876