Learn More
JDM SCIENTIFIC LLC AlteCap™ Switch Cassette 12/96 - External thread Micronic screw caps
Supplier: JDM SCIENTIFIC LLC 404016
AlteCap™ Switch cap driver cassette - 12 channel. Compatible with 96 SBS rack format Micronic external thread screw caps - part numbers MP53825, MP53820, MP53801, MP53804, MP53800, MP53802, MP53809, MP53821, MP53803, MP53806, MP53823, MP53805, MP53822