Learn More
JDM SCIENTIFIC LLC AlteCap™ Solo - External thread Micronic screw caps (24 SBS rack format)
Supplier: JDM SCIENTIFIC LLC 404106
Semi-automated single decapper. Electric capping & decapping. Compatible with 24 SBS rack format Micronic external thread screw caps - part numbers MP535xx.Includes: Manual, stand, USB charging cable & cap driver.