Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell AlphaTec™ BSL4 Orange Protective Suit with Sewn-in Socks
Click to view available options
Size:
2X-Large (Suit), 12 (Socks)
2X-Large (Suit), 13 (Socks)
2X-Large (Suit), 14 (Socks)
Large (Suit), 10 (Socks)
Large (Suit), 11 (Socks)
Large (Suit), 12 (Socks)
Large (Suit), 9 (Socks)
Medium (Suit), 10 (Socks)
Medium (Suit), 7 (Socks)
Medium (Suit), 8 (Socks)
Medium (Suit), 9 (Socks)
Small (Suit), 6 (Socks)
Small (Suit), 7 (Socks)
Small (Suit), 8 (Socks)
X-Large (Suit), 11 (Socks)
X-Large (Suit), 12 (Socks)
X-Large (Suit), 13 (Socks)
X-Large (Suit), 14 (Socks)
Description
- Suitable for Level 4 Biological Safety Laboratories
- Provides protection against chemicals in liquid, aerosol and solid form
- Protects against infectious agents, i.e. bacteria, virus and fungi, and against radioactive particles
- Designed for use with an external air source, which allows longer working hours and provides improved wearer comfort
- Made from a strong yet soft and durable material
Specifications
Color | Orange |
For Use With (Application) | Biosafety Labs Levels 1-4, Centers for Disease Control and Prevention, Life Sciences, Chemical, Mining, Fire and Rescue, Oil and Gas |
Material | Polyester |
Size | 2X-Large (Suit), 14 (Socks) |
Closure Type | Heavy-duty vapor-protective zipper with D-rings |
Disposable | No |
Product Type | Chemical Protective Suits |
Seam Type | Stitched and welded |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction