Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ AlphaTec™ 56-803 Endurosaf™ Aprons
Click to view available options
Size: 34 x 45 in. ; Color: Red
Description
- Strong, tough, 8 to 9 mil die cut Enduro2000™ polyurethane material offers a unique combination of characteristics to outperform neoprene, nitrile, vinyl and other films
- Sealed ties and adjustable neck straps provide a durable, consistent fit for a wide variety of users
- Soft, comfortable, easy to clean and remain flexible in cold temperatures
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | Universal |
Material | Polyurethane |
Color | Red |
For Use With (Application) | Packing fillets and ground meat, Canning, Bottling, Storing and transportation, Equipment maintenance and repair, Plant disinfection and sanitization, Transferring liquids |
Thickness | 8 to 9 mil |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction