Learn More
Ansell™ AlphaTec™ 56-800 Mandiles Endurosaf™ Aprons
Click to view available options
AlphaTec™ 56-800 Mandiles Endurosaf™ Aprons
Description
- Helps keeps workers clean and protected from oil, grease and chemicals
- Endurosaf™ aprons resistant to abrasion, chemicals, oils and fats
- Durable
- Endurosaf™ polyurethane construction is strong, tough and offers tear and puncture resistance yet remains flexible in cold temperatures
- Dielectrically sealed neck and waist straps are easy to keep clean and resist tear out while the neck strap features an adjustable non-corrosive, non-absorbent plastic buckle
- 8 to 9 mil die cut Enduro2000™ polyurethane material that outperforms neoprene, nitrile, vinyl and other films
- Food processing, Life Sciences
Specifications
Material | Polyurethane |
Color | White |
Thickness | 8 to 9 mil |
Includes | 24 Aprons/Cs. |
Dimensions (L x W) | 34 x 50 in. |
Style | AlphaTec 56-800 |