Learn More
Lauda Brinkmann Lp ALPHA IMMERSION CIRCULATOR
Supplier:  Lauda Brinkmann Lp L000630
Encompass_Preferred
Immersion Heating Circulator, Alpha Series, Model Alpha, Digital control interface with simple 3 button operation, for submersing in sample vessels, 1.15 kW Heater, Temperature range of ambient to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 60Hz This item is non returnable*
Catalog No. 50-931-207
Explore available promotions