Learn More
Enzo Life Sciences alpha-Dendrotoxin (140µg). CAS: 74504-53-3
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences BMLKC1670140
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
α-Dendrotoxin is isolated from Dendroaspis angusticeps snake venom, by modification of the procedures of Harvey and Benishin and purified to homogeneity. α-Dendrotoxin blocks Kv1.1 and Kv1.2 channels (IC50= 0.4 to 4 and 1.1 to 12 nM in oocytes respectively, with higher values for mammalian cells). In addition, the toxin was shown to block also Kv1.6 (IC50= 9-25 nM). Purity: ≥98% (HPLC). Solubility: Soluble in any aqueous buffer. Please centrifuge all product preparations at 10000 x g for 5 minutes before reconstitution. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.