Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medline™ Aloetouch™ Nitrile Gloves

Click to view available options
Size:
X-Small
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Aloetouch™ nitrile exam gloves soothe and moisturize hands while providing chemical resistance and barrier protection.

  • Aloe coating helps soothe and moisturize hands
  • Green
  • Full texture for better grip
  • Tested for use with chemotherapy drugs
  • Latex-free
Certifications/Compliance ASTM D 6319
Chemotherapeutic Rated Yes
Applications Healthcare, Industrial
Color Green
Disposable Yes
Exterior Finish Fully Textured
Finger Thickness (English) 5.9 mil
Finger Thickness (Metric) 0.14 mm
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Size Small
Thickness (English) 5.5 mil
Thickness (Metric) 0.13 mm
Model MDS195084
Series Aloetouch™
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less