Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alloxan monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number AC215371000
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
3237-50-1, 50-71-5
C4H4N2O5
MFCD00149399
ZIIHZVKHFWOENY-UHFFFAOYSA-N
3237-50-1, 50-71-5
C4H4N2O5
MFCD00149399
ZIIHZVKHFWOENY-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
Color Beige-Yellow to White
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 24, 500
Merck Index 10, 276
Synonym isovaleraldehyde, 2,4-dinitrophenyl hydrazone, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxybarbituric acid, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, ccris 5957, butanal, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyde 2,4-dinitrophenyl hydrazone, n-e-3-methylbutylideneamino-2,4-dinitroaniline, isovaleraldehyde, 2,4-dinitrophenylhydazone, butyraldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyde dnp, o2aap9f8b6, unii-o2aap9f8b6, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, barbituric acid, 5,5-dihydroxy, 1e-3-methylbutanal 2,4-dinitrophenyl hydrazone #, isovaleraldehyd-2,4-dinitrophenylhydrazone, isovaleraldehyde,4-dinitrophenyl hydrazone
SMILES OC1(O)C(=O)NC(=O)NC1=O
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol) 160.09
PubChem CID 9569524
Formula Weight 160.09
Percent Purity ≥97.5%
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Chemical Name or Material Alloxan monohydrate, 98%
Show More Show Less
Hazard Statement GHS H Statement:

Harmful in contact with skin.

Harmful if swallowed.

Harmful if inhaled.
Precautionary Statement GHS P Statement:

Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN: Wash with plenty of soap and water.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber UW0492000

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit