Learn More
Allegro™ Filter Elements for Ambient Air Pumps
Ambient air pump replacement filter elements help provide safe breathable air to workers in environments with low oxygen levels or compromised air quality.
Supplier:  Allegro™ 970001
 View more versions of this product
Catalog No. 19-060-426
Add to Cart
Description
Description
  • Replacement parts for use with Allegro ambient air pumps
  • Models A-750 and A-740TE
  • Inlet filter and exhaust filter
Specifications
Specifications
Ambient Air Pumps
9700-01
Intake Filter
Product Suggestions
Product Suggestions