Learn More
United Scientific™ All Glass Vacuum Filter Holder Set and Accessories
All Glass Vacuum Filter Holder Set and Accessories used in particulate contamination analysis.
Supplier: United Scientific™ FHSC47
Description
- Used in particulate contamination analysis for filtering aqueous, organic, or corrosive liquids.
- Recommended for HPLC solvent filtration.
- Complete set includes a borosilicate glass funnel, anodized aluminum spring clamp, vacuum base with sintered disc, and ground joint flask.
- Membrane filters are not included.
- Parts are also available separately.
Specifications
Particulate Contamination Analysis | |
Spring Clamp | |
Replacement Spring Clamp, 47 mm |
Borosilicate Glass Funnel and Base, Anodized Aluminum Spring Clamp, and Perforated Silicone Stopper | |
Glass |