Learn More
Sigma Aldrich Fine Chemicals Biosciences Alkane standard mixture for performance tests of GC-systems, 2 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences 682812MLF
Alkane standard mixture has been used as a retention index calibration standard in the metabolic analysis of body fluids of humans and vertebrates using gas chromatography coupled with mass spectrometry (GC-MS).