Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ALK Split FISH Probe
Abnova FISH probes are ensured to provide researchers an effective tool to confirm various genetic amplification, rearrangements, deletions, and aneuploidy.
Supplier: Abnova™ FS0047
Description
- Gene name: ALK
- Gene Alias: CD246, Ki-1, TFG/ALK
- Entrez GeneID: 238
- Gene Description: anaplastic lymphoma receptor tyrosine kinase
Specifications
Store at 4°C in the dark. | |
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. | |
ALK Split FISH Probe | |
200 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only
Spot an opportunity for improvement?
Provide Content Correction