Learn More
Advanced Biomatrix Preformulated alginate 5% concentration
Supplier: Advanced Biomatrix IKA325000503
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alginate 5% is a viscous alginate hydrogel, suitable for both bioprinting and casting. You can use it on its own, mix it with another hydrogel to make the blend ionically crosslinkable or dilute it using our Reconstitution Agent M to achieve your target concentration.
This item is not returnable.
View return policy