Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ Alethia™ Mycoplasma Direct External Controls
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Used as a External Control Materials for DNA amplification assay
Supplier: Meridian Bioscience™ 479890
This item is not returnable.
View return policy
Description
- External controls are used as part of a routine quality control program
- Mycoplasma Direct DNA Amplification Assay Results in less than 1 hour
- Detects Mycoplasma pneumoniae on day 1 of symptoms
- Simple sample preparation from throat swabs
- High sensitivity and specificity
- Quicker initiation of appropriate patient care
- Expanded molecular menu
- No capital expense or service contract required
- Small footprint, easily adaptable to any laboratory
Specifications
CE Marked | |
Yes | |
18 Reactions |
2°C to 8°C | |
Moderate | |
<1 hr. |
Safety and Handling
ShelfLife : 24 Months
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction