Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ Group B Streptococcus Assays
SDP

Definitive and accurate results for improved patient management.

Supplier:  Meridian Bioscience™ 480350

Catalog No. 23-029-006


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Empower your lab with the speed and accuracy of molecular testing for Group B Streptococcus.

  • Provides definitive and accurate results for improved patient management
  • Allows labs to fully comply with CDC guidelines
  • Broth enrichment followed by illumigene™ GBS provides increased sensitivity over culture for the detection of GBS
  • SENSITIVITY 97.4%
  • SPECIFICITY 92.3%

Specifications

Specifications

CE Marked
Streptococci (Group B)
Group B Streptococcus Assay
87081, 87653
Vaginal or Rectal Swab
50 Tests
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit