Learn More
Abbott™ OSTEOMARK™ NTx Urine ELISA
The Osteomark™ NTx Urine ELISA assay is a urinary assay that provides a quantitative measure of the excretion of cross-linked N-telopeptides of type I collagen (NTx) as an indicator of human bone resorption. Monitor response to antiresorptive therapies in postmenopausal women, patients diagnosed with osteoporosis, Pagets disease of bone, or on estrogen-suppressing therapy.
Supplier: Abbott™ X9006
This item is not returnable.
View return policy
Description
- Patient's bone resorption can be tested every three months after initiating treatment
- Results can be a useful complement to bone mineral density tests, which are typically performed every 24 months
- ELISA microwell plate format
- Kits available for urine or serum testing; contain all controls, calibrators, microplates, and reagents
- Room temperature incubation and quantitative results within two hours make this test efficient and effective
- For in vitro diagnostic use
Specifications
High | |
Microwell | |
Osteomark™ NTx Urine ELISA Test Kit | |
96 Tests |
N-terminal telopeptide (NTx) | |
Urine | |
All control, calibrators, microplates and reagents | |
2 hr. |