Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ BinaxNOW™ Malaria Tests
Rapid malaria test cleared by the FDA
Supplier: Abbott™ 665025
This item is not returnable.
View return policy
Description
A rapid and simple diagnostic test that can accurately differentiate a deadly P. falciparum infection from a pan-malarial infection caused by P. vivax, P. ovale, or P. malariae
- For differential diagnosis of a malaria infection
- Just three simple steps, using one reagent
- Reliable results in 15 minutes
- CLIA Complexity: Moderate
- CPT Code: 87899
- Sensitivity: 99.7% (P.f.); 93.5% (P.v.)
- Specificity: 94.2% (P.f.); 99.8% (P.v.)
BNX665025: Malaria Test Kit - US
BNX665010: Positive Control
Specifications
87899 | |
P. vivax Antigen, P. ovale Antigen, P. malariae Antigen | |
Malaria Test Kit (U.S.) | |
25 Tests |
Moderate | |
Whole Blood | |
Malaria Test Kit (U.S.), with built in control | |
15 min. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction