Learn More
MilliporeSigma™ Supelco™ Aldrich™ Chromatography Sprayer
Provide a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
Supplier: MilliporeSigma™ Supelco™ Z5297451EA
Description
Adjustable spray pattern using thumb on vent hole. Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely. Threaded SafetyBarbs™ prevent accidental breakage during tubing installation or removal. Uses low-pressure gas or air.Specifications
250 mL | |
TLC | |
Aldrich |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC