Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Blood Typing Anti-Sera, Anti-A, Anti-B and Anti Rh
Use for blood typing of real human blood. Each vial contains enough antisera for 75 students. Refrigeration recommended until used. Materials from human-derived cell lines were found to be negative in tests for Hepatitis B, HIV and EBV.
$39.00 - $79.00
Specifications
Product Type | Blood Typing Anti-Sera |
---|---|
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | Use for blood typing of real human blood |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S07802
|
Aldon Corporation
IS3170 |
Anti-a and anti-b |
Each for $39.00
|
|
|||||
S07803
|
Aldon Corporation
IS3171 |
Type a, b and rh |
Each for $79.00
|
|
|||||
S07804
|
Aldon Corporation
IS3172 |
Anti-rh |
Each for $39.25
|
|
|||||
Description
- Available in 5mL vials that contain enough antisera for 75 students
- Freeze-dried samples; can be kept without refrigeration
- Shows negative results for Hepatitis B, HIV and EBV when tested with human-derived cell lines
Specifications
Blood Typing Anti-Sera | |
7 to 14 | |
Use for blood typing of real human blood | |
3 x .0.5 x 0.5 in. |
White | |
13 to 20 | |
1 | |
75 Students |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title