Learn More
Alcohols in Water, WS Proficiency Testing Material, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Supplier: MilliporeSigma Supelco PE13121.2ML
Specifications
1.2 mL | |
1-Pentanol 5-50 mg/L; 1-Propanol 2.5-20 mg/L; 2-Butanol 5-50 mg/L; 4-Methyl-2-Pentanone (MIBK) 1-50 mg/L; Ethanol 5-50 mg/L; Iso-Butanol 5-50 mg/L; Isopropyl Alcohol (2-Propanol) 2.5-50 mg/L; Methanol 5-100 mg/L; n-Butyl Alcohol 5-50 mg/L; tert-Butyl Alcohol 5-50 mg/L; Tetrahydrofuran (THF) 5-50 mg/L |