Learn More
Cole-Parmer Cole-Parmer Alcohol Lamp with Snuffer Cap and Extra Wick, 135 mL Capacity
Supplier: Cole-Parmer 3630020
Alcohol lamp is ideal for laboratories where there is no gas source. It is made of nickel-plated brass. Wicks are 6" to 7" high. Optional two-way lamp stand .The distance from the lamp stand to sample is 4" or 5", depending on orientation of stand. Lamp includes snuffer cap and one extra wick.