Learn More
Cole-Parmer DWK Life Sciences (Wheaton) 237070 Alcohol Burner, 100 ml Capacity, 4" H x 3" Dia
Supplier: Cole-Parmer 4858584
Manufactured from soda-lime glass, this burner is designed for use with isopropyl or denatured ethyl alcohol. The low-heat flame is well suited for microscopy purposes.