Learn More
Andwin Scientific ALCOHOL BURNER
SDP
Supplier:  Andwin Scientific 17805005
Encompass_Preferred
The glass alcohol burner by Grobet features a clear glass base that allows convenient and easy viewing of the fuel level and eliminates the necessity of stopping during procedures to check the level. Its wick height is modifiable with no difficulty. The metal cap is fitted with a sealing gasket to avoid leakage. This product includes a glass reservoir, wick holder, gasket, wick, and storage cap.
Catalog No. NC9360811
Explore available promotions