Learn More
Electron Microscopy Sciences Alcohol Burner, 100 ml
SDP
Supplier:  Electron Microscopy Sciences 6421301
Encompass_Preferred
This burner offers a low heat flame, which is well suited for microscopy purposes. For use with isopropyl and denatured ethyl alcohol. The unit is supplied with a ground glass stopper. Made from soda lime glass. Size is 100 ml and measures: 75 mm in diameter and 100 mm high.
Catalog No. 50-298-52