Learn More
Newcomer Supply Alcian Blue/PAS Control Slides, 98/set
Supplier: Newcomer Supply 4022B
Newcomer Supply Alcian Blue/PAS Control Slides are for the positive histochemical staining and differentiation of acidic epithelial mucins (sialomucin, sulfomucin), stromal (mesenchymal) mucin, neutral mucins and glycogen. Alcian Blue/PAS Control Slides contain sections of positive staining small intestine.