Learn More
Spectrum™ Chemical Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade
SDP
A3611, 9048-46-8
$200.02 - $2681.40
Specifications
Packaging Poly Bottle
Form Lyophilized
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-600-351
SDP
View Documents
Spectrum™ Chemical
A361125GM
25 g Poly Bottle
Each for $200.02
Add to Cart
18-600-352
SDP
View Documents
Spectrum™ Chemical
A3611100GM
100 g Poly Bottle
Each for $579.88
Add to Cart
18-600-353
SDP
View Documents
Spectrum™ Chemical
A36111KGBL
1 kg Poly Bottle
Each for $2,681.40
Add to Cart
Description
Description
Spectrum™ Chemical Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade is a serum albumin protein that comes from cows. It is also known as Bovine serum albumin and usually used as a protein concentration standard in laboratory experiments. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications
Specifications
Poly Bottle
Lyophilized