Learn More
AirClean™ Systems HEPA Filters
![SDP](/content/dam/fishersci/glyphs/sdp.png)
High efficiency particulate air (HEPA) filter, AirClean™ systems HEPA filters can be used with ductless fume hoods and drying cabinets.
Supplier: AirClean™ Systems ACFHEPA56
Description
- High efficiency particulate air (HEPA) filters with 99.997% efficiency rating for collection at 0.3 microns
- Used for containment of particles - toxic powders, asbestos, lead, fingerprint powders
Specifications
HEPA | |
Horizontal Laminar Flow Workstation (AC5000HLF and UV) | |
Filter |