Learn More
Air Systems™ International Remote Replacement Cylinders

Disposable replacement cylinders for accurate calibration
$75.90 - $308.00
Specifications
Applications | For calibration |
---|---|
For Use With (Equipment) | For remote airline system alarms |
Product Type | Replacement Cylinder |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
19-806-329
![]() ![]() |
Air Systems™
BBG20103 |
27.2 gal. | 103 L | 103 L Replacement CO cylinder |
Each for $308.00
|
|
|||
19-806-326
![]() ![]() |
Air Systems™
BBG00 |
4.49 gal. | 17 L | 17 L Replacement Zero Reference Cylinder |
Each for $75.90
|
|
|||
Specifications
For calibration | |
For remote airline system alarms | |
Replacement Cylinder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.