Learn More
Cole-Parmer Air Cadet Vacuum/Pressure Pump, Diaphragm, Single Head, 0.6 cfm, 115 VAC
Supplier: Cole-Parmer 0753240
The unique cavity configuration of this vacuum and pressure pump minimizes dead space and air entrapment. It features an all-plastic, convoluted diaphragm design to handle many vacuum and pressure applications and reduce stress, wear and heat build-up. The pumps are available with a standard 115 or 230 VAC motor or, portable 12 VDC one. The standard AC motors are service-free, brushless with sealed ball bearings, and thermal overload protection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.